Learn More
CELLTREAT Scientific Products 200µL Gel Loading Tips, Round, Racked, Non-sterile
SDP
Catalog No. 5000144328
Encompass_Preferred
Round, fine point is ideal for loading samples into electrophoresis gelsUniversal fit is compatible with major brands of pipettesRacked tips follow standard spacing for use with multichannel pipettes
Catalog No. 50-001-44328 Supplier CELLTREAT Scientific Products Supplier No. 229028
Round, fine point is ideal for loading samples into electrophoresis gelsUniversal fit is compatible with major brands of pipettesRacked tips follow standard spacing for use with multichannel pipettes