Learn More
Analytical Sales & Services 200µL PEEK Sample Loop
Catalog No. 5000804914
Encompass
200µL PEEK Sample Loop for High Pressure Automatic Switching Valves and 9125, 9725 and 9010 Valves\n\nQty/Pkg: EACH
Catalog No. 50-008-04914 Supplier Analytical Sales & Services Supplier No. 490550254
200µL PEEK Sample Loop for High Pressure Automatic Switching Valves and 9125, 9725 and 9010 Valves\n\nQty/Pkg: EACH