Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Axygen™ 200 μL Universal Pipetter Tips: 200 μL, Beveled, Reference

Catalog No. 14222819 Shop All Axygen Products
Change view
Click to view available options
Volume (Metric):
1 to 200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
14-222-819 1 to 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 14-222-819 Supplier Axygen™ Supplier No. TR222CSTKS
This item is not returnable. View return policy

Designed to fit a wide variety of single and multi-channel pipettes

Axygen™ Universal Tips are designed to fit a wide variety of single and multi-channel pipettors. These polypropylene tips are nonpyrogenic and certified to be RNase-/DNase-free. They are available in Maxymum Recovery, beveled, Eppendorf-style, non-sterile, sterile, non-filtered, and other features for specialized applications. Flexible walls and a series of internal sealing rings ensure a secure fit with less force required to load and eject.
(This is the suggested replacement for T-205-WB-C-L)
  • These polypropylene tips are nonpyrogenic and certified to be RNase-/DNase-free.
  • Nonpyrogenic
  • Wide-Bore
  • TRUSTED_SUSTAINABILITY

    Specifications

    Volume (Metric) 1 to 200 μL
    Filtered Non-filtered
    Sterility Sterile
    Color Clear
    Autoclavable Autoclavable
    Certifications/Compliance EN ISO 13485 compliant, BSE/TSE compliant, Non-Pyrogenic, USP Class VI tested, Human DNA Free, DNase/RNase Free, Quality Control Tested
    Clean Claims Maxymum Recovery, RNase-free, DNase-free, Nonpyrogenic, Human DNA-free, ATP-free
    Format Refill
    Tip Style Beveled
    Quantity 480/Pk., 4,800/Cs.
    For Use With (Equipment) Universal Fit
    Material Polypropylene
    Show More Show Less
    WARNING: Cancer - www.P65Warnings.ca.gov
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.