Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Axygen™ 200μL Universal Pipetter Tips: 200μL, Beveled

Catalog No. 14222722 Shop All Axygen Products
Change view
Click to view available options
Volume (Metric):
1 to 200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
14-222-722 1 to 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14-222-722 Supplier Axygen™ Supplier No. T200YR
Add to Cart
Add to Cart

Designed to fit a wide variety of single and multi-channel pipettes

  • Optimized tip bevel reduces sample hang-up, improves pipetting accuracy
  • Maxymum recovery allows complete retrieval of precious samples, saves costly reagents
  • Wide bore tips of orifice size: 0.062 in. (1.52 mm) makes it ideal for pipetting viscous materials, genomic DNA or delicate cell cultures
  • Graduation marks allow a visual confirmation of pipettor draw volume
  • Presterilized tips are e-beam irradiated to prevent product degradation
  • Clean-room manufacturing environment certifies products safe from RNase, DNase and pyrogen safe

TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 1 to 200 μL
Filtered Non-filtered
Sterility Non-sterile
Color Yellow
Autoclavable Autoclavable
Model T-200-Y-R
Clean Claims RNase/DNase-free, Nonpyrogenic
Format Rack
Tip Style Beveled
Quantity 4800 Tips/Cs.
Product Type Pipette Tips
For Use With (Equipment) Universal Fit
Graduated Yes
Product Line Maxymum Recovery
Material Polypropylene
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.