Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Axygen™ 200 μL Universal Pipetter Tips: 200 μL, Wide Bore

Catalog No. 14222729 Shop All Axygen Products
Change view
Click to view available options
Volume (Metric):
1 to 200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
14-222-729 1 to 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14-222-729 Supplier Axygen™ Supplier No. T205WBCR

missing translation for 'validMainframeMsg'

Designed to fit a wide variety of single and multi-channel pipettes

Axygen™ Universal Tips are designed to fit a wide variety of single and multi-channel pipettors. These polypropylene tips are nonpyrogenic and certified to be RNase-/DNase-free. They are available in Maxymum Recovery, beveled, Eppendorf-style, non-sterile, sterile, non-filtered, and other features for specialized applications. Flexible walls and a series of internal sealing rings ensure a secure fit with less force required to load and eject.
(This is the suggested replacement for T-205-WB-C-L)
  • These polypropylene tips are nonpyrogenic and certified to be RNase-/DNase-free.
  • Nonpyrogenic
  • Wide-Bore
  • '
    TRUSTED_SUSTAINABILITY

    Specifications

    Volume (Metric) 1 to 200 μL
    Filtered Non-filtered
    Sterility Non-sterile
    Color Clear
    Rack Color Clear
    Autoclavable Autoclavable
    Model T-205-WB-C-R
    Certifications/Compliance ISO 9001 Standard compliant, ISO 13485 Standard compliant, BSE/TSE compliant, USP Class VI tested, Non-Pyrogenic, Human DNA Free, DNase/RNase Free, Quality Control Tested (Visual Inspection - Pass, Dimensional Inspection)
    Clean Claims DNase, RNase, and Pyrogen free
    Format Rack
    Tip Style Genomic/Wide Orifice
    Quantity 960/Pk., 4800/Cs.
    Product Type Pipette Tips
    For Use With (Equipment) Universal Fit
    Product Line Maxymum Recovery
    Material Polypropylene
    Show More Show Less
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.