Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Troemner™ 200 mg Analytical Precision UltraClass Weight with Traceable Certificate

Catalog No. 02218093 Shop All Troemner Products
Change view
Click to view available options
Traceable Certificate
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
02-218-093 Traceable Certificate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 02-218-093 Supplier Troemner™ Supplier No. 30390213
Add to Cart
Add to Cart

Combine high precision with the advantage of two-piece construction. UltraClass weights, exclusive to Troemner™ are the most precise two-piece weights available with weight tolerances that equal ANSI/ASTM E617 Class 0 or OIML R 111 Class E2 tolerances.

  • Used in extremely stable environments
  • One-piece construction with no adjusting cavity with one end turned up for easy handling
  • Supplied in a precision-fit, high quality polycarbonate case
  • Includes a Traceable Certificate
TRUSTED_SUSTAINABILITY

Specifications

Product Type One-Piece
Certifications/Compliance Traceable Certificate
Classification UltraClass
Material Stainless Steel
Case Material Polycarbonate
Density 7.95 g/cm3 at 20°C
Mass 200 mg
Tolerance 0.005 mg
Length (Metric) 5.08 cm
Height (Metric) Case 6.35 cm
For Use With (Application) Reference standard calibration
For Use With (Equipment) Calibration of high-precision analytical balances with readability as low as 0.01 mg
Includes Sheet Weight, Polycarbonate Case
Quantity Each
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.