Learn More
Antron Engineering Syringe 200ML Stainless Steel PRO-7600
SDP
Catalog No. 50871816
Encompass_Preferred
These syringes are intended for high-pressure applications and offer good resistance to most aggressive liquids. Syringe barrels and plungers are made of type 316 stainless steel. Syringes are leak-free to pressures up to 750psi, except for 8mL syringes that are leak-free to pressures up to 1500psi. They fit most syringe pumps. Autoclavable.
Catalog No. 50-871-816 Supplier Antron Engineering Supplier No. PRO7600LL
These syringes are intended for high-pressure applications and offer good resistance to most aggressive liquids. Syringe barrels and plungers are made of type 316 stainless steel. Syringes are leak-free to pressures up to 750psi, except for 8mL syringes that are leak-free to pressures up to 1500psi. They fit most syringe pumps. Autoclavable.