Learn More
Omega Biotek Inc 200 PACK OF COLUMNS
Catalog No. NC0567415
Encompass
HiBind DNA Columns are silica glass fiber columns that can bind up to 100 ug of genomic DNA or 35 ug of plasmid DNA per prep. Columns feature an attached lid and a vacuum luer for processing with vacuum manifolds and by centrifugation. Omega Bio-tek Columns are compatible with the following Qiagen Kits: Qiaprep Spin Mini, QiaQuick Gel Extraction, QiaQuick PCR Clean-up, Dneasy Blood & Tissue, Dneasy Plant DNA. 200 preps per each.
Catalog No. NC0567415 Supplier Omega Biotek Inc Supplier No. DNACOL02
HiBind DNA Columns are silica glass fiber columns that can bind up to 100 ug of genomic DNA or 35 ug of plasmid DNA per prep. Columns feature an attached lid and a vacuum luer for processing with vacuum manifolds and by centrifugation. Omega Bio-tek Columns are compatible with the following Qiagen Kits: Qiaprep Spin Mini, QiaQuick Gel Extraction, QiaQuick PCR Clean-up, Dneasy Blood & Tissue, Dneasy Plant DNA. 200 preps per each.