Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

200 Proof Alcohol for Extraction CDA-12A (Ethanol and n-Heptane), Reagents

Catalog No. 01337530
Change view
Click to view available options
Quantity:
20 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-337-530 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-337-530 Supplier Reagents Holdings Llc Supplier No. C212080020E6
Add to Cart
Add to Cart

200 Proof Alcohol for Extraction CDA-12A (Ethanol and n-Heptane), Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 64-17-5 , 142-82-5
Molecular Formula C2H6O
Molecular Weight (g/mol) 46.07
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
Synonym Ethanol, Ethyl alcohol, Methylcarbinol
IUPAC Name ethanol
SMILES CCO

Specifications

Melting Point -55°C
Color Colorless
Quantity 20 L
Packaging Steel Pail
Physical Form Liquid
Specific Gravity 0.7974
Boiling Point 78°C
Percent Purity Min. 99.5%
Grade Reagent
Chemical Name or Material 200 Proof Alcohol for Extraction CDA-12A (Ethanol + n-Heptane)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.