Learn More
Chemglass Life Sciences Chemglass 200 μL automation conductive filter tips for Tecan - sterile, 96/rack, 10 racks/pack (CLS-3499A-092)
SDP
Catalog No. 502733253
Encompass_Preferred
Chemglass 200 μL automation conductive filter tips are engineered for high-throughput liquid handling on Tecan systems. Made from medical-grade polypropylene and electron-beam sterilized, the tips include aerosol-resistant filters to protect samples and instrumentation. Packaging is 96 tips per rack, 10 racks per pack, and 50 racks per case.
  • 200 μL nominal volume.
  • Designed for Tecan automated platforms.
  • Medical-grade polypropylene construction.
  • Electron beam sterilized and DNase/RNase free.
  • Aerosol-resistant filter to minimize contamination.
  • 96 tips per rack; 10 racks per pack; 50 racks per case.
  • Manufactured in a 100,000-grade clean room.
Catalog No. 50-273-3253 Supplier Chemglass Life Sciences Supplier No. CLS3499A092
Add to Cart
Add to Cart
Chemglass 200 μL automation conductive filter tips are engineered for high-throughput liquid handling on Tecan systems. Made from medical-grade polypropylene and electron-beam sterilized, the tips include aerosol-resistant filters to protect samples and instrumentation. Packaging is 96 tips per rack, 10 racks per pack, and 50 racks per case.
  • 200 μL nominal volume.
  • Designed for Tecan automated platforms.
  • Medical-grade polypropylene construction.
  • Electron beam sterilized and DNase/RNase free.
  • Aerosol-resistant filter to minimize contamination.
  • 96 tips per rack; 10 racks per pack; 50 racks per case.
  • Manufactured in a 100,000-grade clean room.