Learn More
USA Scientific Inc 200 uL flat gel tip, 0.37 mm
Catalog No. 502745280
Encompass
200 uL flat sequencing gel tip, 0.37 mm OD orifice, sterilized, 96 tips/rack, 2 racks/pack (192 tips)
Catalog No. 50-274-5280 Supplier USA Scientific Inc Supplier No. 10224610
200 uL flat sequencing gel tip, 0.37 mm OD orifice, sterilized, 96 tips/rack, 2 racks/pack (192 tips)