Learn More
USA Scientific Inc 200 uL round gel tip, bulk
Catalog No. 502745167
Encompass
200 uL Gel-Saver round gel loading tip, 0.6 mm OD orifice, bulk (1000 tips)
Catalog No. 50-274-5167 Supplier USA Scientific Inc Supplier No. 10220000
200 uL Gel-Saver round gel loading tip, 0.6 mm OD orifice, bulk (1000 tips)