Learn More
Ace Glass, Inc. 10mm 46cm stainless steel stir shaft, polished, removable button type, 20cm flat section
SDP
Catalog No. 509558206
Encompass
10mm O.D. 46cm stainless steel polished stir shaft, drilled type, with removable button. Stir Shaft also has a 20cm flat section for use with agitators secured by set screws. For use with Trubore and PTFE bearings.
Catalog No. 50-955-8206 Supplier Ace Glass, Inc. Supplier No. 807404
10mm O.D. 46cm stainless steel polished stir shaft, drilled type, with removable button. Stir Shaft also has a 20cm flat section for use with agitators secured by set screws. For use with Trubore and PTFE bearings.