Learn More
Biotium GloMelt™ Thermal Shift Protein Stability Kit, with ROX, 2000 assays
Catalog No. NC1618224
Encompass_Preferred
GloMelt™ dye can be used to detect protein unfolding or measure thermal stability by performing a thermal shift assay, also called Protein Thermal Shift™, differential scanning fluorimetry, or Thermofluor assay.
Catalog No. NC1618224 Supplier Biotium Supplier No. 330221
GloMelt™ dye can be used to detect protein unfolding or measure thermal stability by performing a thermal shift assay, also called Protein Thermal Shift™, differential scanning fluorimetry, or Thermofluor assay.