Learn More
Electron Microscopy Sciences Fine 2000 Mesh 25x25 mm Square, Nickel
SDP
Catalog No. 5025926
Encompass_Preferred
These fine mesh grids are carefully checked. They are suitable for the low magnification range of a TEM. In a square mesh 25mm.
Material: Nickel
Catalog No. 50-259-26 Supplier Electron Microscopy Sciences Supplier No. 7952620
These fine mesh grids are carefully checked. They are suitable for the low magnification range of a TEM. In a square mesh 25mm.
Material: Nickel