Learn More
Electron Microscopy Sciences Fine 2000 Mesh on Sandwich Grid, Copper
SDP
Catalog No. 5025925
Encompass_Preferred
These fine mesh grids are carefully checked. They are suitable for the low magnification range of a TEM. Sandwich in a folding 3.05mm mesh grid.
Material: Copper
Catalog No. 50-259-25 Supplier Electron Microscopy Sciences Supplier No. 7952502
These fine mesh grids are carefully checked. They are suitable for the low magnification range of a TEM. Sandwich in a folding 3.05mm mesh grid.
Material: Copper