Learn More
Chemglass Life Sciences 2, 000mL Jacketed Reaction Beaker
SDP
Catalog No. CG110306
Encompass_Preferred
2,000mL Jacketed Reaction Beaker. Designed for use in temperature controlled chemical and biological reactions. Bottom interior surface is conical in shape to help eliminate spinout when used with CG-2005 egg shaped spinbars. Hose connections have an O.D. of 10mm at the largest serration.
Catalog No. CG110306 Supplier Chemglass Life Sciences Supplier No. CG110306
2,000mL Jacketed Reaction Beaker. Designed for use in temperature controlled chemical and biological reactions. Bottom interior surface is conical in shape to help eliminate spinout when used with CG-2005 egg shaped spinbars. Hose connections have an O.D. of 10mm at the largest serration.