Learn More
Brookfield Engineer Labs B Mineral Oil Fluids, B200000
Catalog No. NC0933068
Encompass
Brookfield Viscosity Standards are accurate to ±1% of the stated viscosity and are certified by methods traceable to the United States National Institute of Standards and Technology (NIST). The selection of one or two fluids will normally provide sufficient measurement points to verify the calibration of your instrument. All fluids are supplied in 1/2 liter (1 pint) containers complete with a certificate of calibration.
Catalog No. NC0933068 Supplier Brookfield Engineer Labs Supplier No. B200008
Brookfield Viscosity Standards are accurate to ±1% of the stated viscosity and are certified by methods traceable to the United States National Institute of Standards and Technology (NIST). The selection of one or two fluids will normally provide sufficient measurement points to verify the calibration of your instrument. All fluids are supplied in 1/2 liter (1 pint) containers complete with a certificate of calibration.