Learn More
Ace Glass, Inc. 2000mL graduated cylindrical funnel, 24/40 top, 29/42 bottom
SDP
Supplier:  Ace Glass, Inc. 726870
Encompass_Preferred
2000mL graduated cylindrical funnel with 24/40 joint at top and 29/42 joint at bottom. Features include drip stem, and double scale graduations calibrated from stopcock. Drip stem does not extend beyond the joint, thereby eliminating a possible source of breakage. Includes PTFE stopcock plug.
Catalog No. 50-974-739