Learn More
Ace Glass, Inc. 2000mL 45/50 separatory funnel with polyethylene stopper
SDP
Catalog No. 50974693
Encompass_Preferred
2000ml separatory funnel with polyethylene stopper. Outer ground joint at top is a 45/50 Rodaviss joint. Stopper can be secured in top joint via cap and O-ring for a leak-tight seal.
Catalog No. 50-974-693 Supplier Ace Glass, Inc. Supplier No. 723121
2000ml separatory funnel with polyethylene stopper. Outer ground joint at top is a 45/50 Rodaviss joint. Stopper can be secured in top joint via cap and O-ring for a leak-tight seal.