Learn More
Chemglass Life Sciences 2000mL Addition Funnel, 29/42 Joint Size, 6mm PTFE Stpk, 535mm OAH
SDP
Catalog No. CG170209
Encompass_Preferred
2000mL Addition Funnel, 29/42 Joint Size, 6mm PTFE Stopcock, 535mm OAH. Cylindrical style addition funnel with a standard taper top outer joint and lower inner joint, both being the same size. With pressure equalizing arm and PTFE stopcock. For replacement stoppers see CG-3000.
Catalog No. CG170209 Supplier Chemglass Life Sciences Supplier No. CG170209
2000mL Addition Funnel, 29/42 Joint Size, 6mm PTFE Stopcock, 535mm OAH. Cylindrical style addition funnel with a standard taper top outer joint and lower inner joint, both being the same size. With pressure equalizing arm and PTFE stopcock. For replacement stoppers see CG-3000.