Learn More
Chemglass Life Sciences 2000mL Addition Funnel, Graduated, 29/42 Joint Size, 6mm PTFE Stpk, 535mm OAH
SDP
Catalog No. CG171809
Encompass_Preferred
2000mL Addition Funnel, Graduated, 29/42 Joint Size, 6mm PTFE Stpk, 535mm OAH. Cylindrical style addition funnel similar to CG-1716, but with PTFE stopcocks. Funnels with 14/20 and 19/22 joints are supplied complete with a stopper. For replacement stoppers see CG-3000.
Catalog No. CG171809 Supplier Chemglass Life Sciences Supplier No. CG171809
2000mL Addition Funnel, Graduated, 29/42 Joint Size, 6mm PTFE Stpk, 535mm OAH. Cylindrical style addition funnel similar to CG-1716, but with PTFE stopcocks. Funnels with 14/20 and 19/22 joints are supplied complete with a stopper. For replacement stoppers see CG-3000.