Learn More
Chemglass Life Sciences 2,000mL Bottle Assembly, 2 Port, Sterile
SDP
Catalog No. 501532080
Encompass
2,000mL Bottle Assembly, 2 Port, Sterile. Bio-Simplex sterile bottle assembly systems are available in a range of standard sizes, from 60mL to 2L. Each system includes a PETG bottle and a 2-port EZ Top Closure Assembly consisting of a 0.2 m vent filter, an 18 inch length of tubing with a plug end, a notched dip tube that extends to the bottom of the bottle and a polyethylene cap. The EZ Top assembly is made from C-Flex pharmaceutical grade thermoplastic elastomer (TPE) resin. The entire system is designed to ensure an unobstructed fluid path and extremely low levels of extractables. Each unit is double poly bagged and gamma irradiated at 25-35 kGy.
Catalog No. 50-153-2080 Supplier Chemglass Life Sciences Supplier No. CLS117002LM
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.
2,000mL Bottle Assembly, 2 Port, Sterile. Bio-Simplex sterile bottle assembly systems are available in a range of standard sizes, from 60mL to 2L. Each system includes a PETG bottle and a 2-port EZ Top Closure Assembly consisting of a 0.2 m vent filter, an 18 inch length of tubing with a plug end, a notched dip tube that extends to the bottom of the bottle and a polyethylene cap. The EZ Top assembly is made from C-Flex pharmaceutical grade thermoplastic elastomer (TPE) resin. The entire system is designed to ensure an unobstructed fluid path and extremely low levels of extractables. Each unit is double poly bagged and gamma irradiated at 25-35 kGy.