Learn More
Chemglass Life Sciences ChemRxnHub Jacketed Process System, 2 L, with Manifold, Stand, and Digital Overhead Stirrer (Chemglass CG-1949-X-2DM)
SDP
Catalog No. NC2195337
Encompass_Preferred
The ChemRxnHub jacketed 2 L process system is a complete benchtop reactor package designed for laboratory synthesis, process development, and small-scale scale-up. It combines a 2,000 mL jacketed reaction vessel with a benchtop support stand, a digital overhead stirrer, a full set of glassware, and an integrated manifold/drain system to simplify temperature control, agitation, and material transfer.
  • Jacketed 2,000 mL reaction vessel for controlled heating and cooling.
  • Integrated manifold and drain system for convenient transfer and sampling.
  • Benchtop stand and secure clamping for stable operation.
  • Digital overhead stirrer for precise mixing and speed control.
  • Complete glassware set compatible with standard laboratory fittings.
  • Suitable for synthesis, reaction screening, and small-scale process development.
This item is not returnable. View return policy
Catalog No. NC2195337 Supplier Chemglass Life Sciences Supplier No. CG1949X2DM
Add to Cart
Add to Cart
Request a Quote
The ChemRxnHub jacketed 2 L process system is a complete benchtop reactor package designed for laboratory synthesis, process development, and small-scale scale-up. It combines a 2,000 mL jacketed reaction vessel with a benchtop support stand, a digital overhead stirrer, a full set of glassware, and an integrated manifold/drain system to simplify temperature control, agitation, and material transfer.
  • Jacketed 2,000 mL reaction vessel for controlled heating and cooling.
  • Integrated manifold and drain system for convenient transfer and sampling.
  • Benchtop stand and secure clamping for stable operation.
  • Digital overhead stirrer for precise mixing and speed control.
  • Complete glassware set compatible with standard laboratory fittings.
  • Suitable for synthesis, reaction screening, and small-scale process development.