Learn More
Chemglass Life Sciences Clamp, 2000mL Flask Holder
SDP
Catalog No. 501214926
Encompass
Clamp, 2000mL Flask Holder. Stainless steel flask clamps are equipped with stainless steel springs to securely hold flask. Clamps are sold 1 per case. The CLS-2446-28L is designed for Fernbach style flasks.
Catalog No. 50-121-4926 Supplier Chemglass Life Sciences Supplier No. CLS24462L
Clamp, 2000mL Flask Holder. Stainless steel flask clamps are equipped with stainless steel springs to securely hold flask. Clamps are sold 1 per case. The CLS-2446-28L is designed for Fernbach style flasks.