Learn More
Chemglass Life Sciences Chemglass Cylinder, Class A, 2000 mL Pyrex Graduated Cylinder, Double Metric Scale (CG-8240-2L)
SDP
Catalog No. NC1963993
Encompass_Preferred
Class A 2000 mL graduated cylinder made from PYREX glass with a double metric scale and calibrated "To Deliver" (TD) for accurate liquid measurement and dispensing. The cylinder includes a shock-absorbing polyethylene bumper guard for added protection and is intended for routine laboratory measurement tasks.
  • Class A accuracy for precise measurement.
  • Double metric scale numbered up and down for easy reading.
  • PYREX glass construction resists chemical attack and thermal shock.
  • Calibrated to deliver (TD) for dispensing accuracy.
  • Shock-absorbing polyethylene bumper guard included to protect the glass.
  • Suitable for routine laboratory liquid measurement and dispensing.
Catalog No. NC1963993 Supplier Chemglass Life Sciences Supplier No. CG82402L
Class A 2000 mL graduated cylinder made from PYREX glass with a double metric scale and calibrated "To Deliver" (TD) for accurate liquid measurement and dispensing. The cylinder includes a shock-absorbing polyethylene bumper guard for added protection and is intended for routine laboratory measurement tasks.
  • Class A accuracy for precise measurement.
  • Double metric scale numbered up and down for easy reading.
  • PYREX glass construction resists chemical attack and thermal shock.
  • Calibrated to deliver (TD) for dispensing accuracy.
  • Shock-absorbing polyethylene bumper guard included to protect the glass.
  • Suitable for routine laboratory liquid measurement and dispensing.