Learn More
Ace Glass, Inc. 2000mL hemispherical fabric mantle, Glas-Col, 170mm max flask diameter, 3.60in inside depth, 8.70in OD, 4.60in height, 500 watts, CSA rated
SDP
Catalog No. 50980643
Encompass
2000mL hemispherical fabric Glas-col mantle, adaptable to unusual shapes and sizes, and will withstand 450 Deg.C. Mantle has a separable 2-wire 4 cord with locking connector. cULus rated. CSA certified.
Catalog No. 50-980-643 Supplier Ace Glass, Inc. Supplier No. 1203521
2000mL hemispherical fabric Glas-col mantle, adaptable to unusual shapes and sizes, and will withstand 450 Deg.C. Mantle has a separable 2-wire 4 cord with locking connector. cULus rated. CSA certified.