Learn More
MAGIC BIOPROCESSING MAGic™Accio Pilot 2000
Catalog No. NC3724030
Encompass_Preferred
MAGic™Accio PILOT is designed to support development-phase and medium-scale purification of nucleic acids, peptides, proteins, antibodies, and viruses from both clarified and unclarified cell culture mixtures. Capable of processing volumes from 5 mL to several liters, it works seamlessly with 20 µL to 250 mL of settled MAGic™ Beads. Used with the MAGic™Accio PILOT 50, 500, or 2000 units, it enables rapid, durable manual separation in both serial and parallel configurations. By combining units, users can isolate low-expressing targets from large volumes and concentrate them into smaller vessels, streamlining workflows and boosting yield.
Catalog No. NC3724030 Supplier MAGIC BIOPROCESSING Supplier No. 2400
Add to Cart
Add to Cart
Request a Quote
MAGic™Accio PILOT is designed to support development-phase and medium-scale purification of nucleic acids, peptides, proteins, antibodies, and viruses from both clarified and unclarified cell culture mixtures. Capable of processing volumes from 5 mL to several liters, it works seamlessly with 20 µL to 250 mL of settled MAGic™ Beads. Used with the MAGic™Accio PILOT 50, 500, or 2000 units, it enables rapid, durable manual separation in both serial and parallel configurations. By combining units, users can isolate low-expressing targets from large volumes and concentrate them into smaller vessels, streamlining workflows and boosting yield.