Learn More
CELLTREAT Scientific Products 2L Square Media Bottle (PETG), Tamper Evident, Bulk, Sterile
SDP
Catalog No. NC3733974
Encompass_Preferred
Lightweight plastic material is easier and safer to handle than glass
Square media bottle shape conserves space in refrigerated storage areas
PET bottles feature clearly printed graduations and defined marking area
PETG bottles feature molded graduations
All bottles are sterile, and available individually wrapped or tray packed
Catalog No. NC3733974 Supplier CELLTREAT Scientific Products Supplier No. 230766TE
Lightweight plastic material is easier and safer to handle than glass
Square media bottle shape conserves space in refrigerated storage areas
PET bottles feature clearly printed graduations and defined marking area
PETG bottles feature molded graduations
All bottles are sterile, and available individually wrapped or tray packed