Learn More
Chemglass Life Sciences 2000mL Single Neck Evaporating Flask, 29/42 Outer Joint, Recovery Flask
SDP
Catalog No. CG151239
Encompass_Preferred
2000mL Single Neck Evaporating Flask, 29/42 Outer Joint, Recovery Flask. Evaporating flask for use with rotary evaporators. Flasks have a HEAVY WALL for vacuum applications and are balanced to prevent run-out during rotation. For Plastic Coated flasks, see CG-1512-P.
Catalog No. CG151239 Supplier Chemglass Life Sciences Supplier No. CG151239
2000mL Single Neck Evaporating Flask, 29/42 Outer Joint, Recovery Flask. Evaporating flask for use with rotary evaporators. Flasks have a HEAVY WALL for vacuum applications and are balanced to prevent run-out during rotation. For Plastic Coated flasks, see CG-1512-P.