Learn More
Bellco Glass, Inc. 2000mL Roller Bottle
SDP
Catalog No. 502330209
Encompass_Preferred
Available in two sizes 1000mL volume with 490cm2 of surface area and 2000mL volume with 850cm2 of surface area treated or nontreated versions. Expanded Surface Roller Bottle increases surface area for cell culture.Specifications Large knurled cap for easy grip Vented or nonvented cap for open or closed systems Manufactured from medical grade virgin polystyrene Tissue Culture Treated Roller Bottles have uniform hydrophilic surface ideal for consistent cell attachment Easytoread black printed graduations Vent caps feature 0.22µm hydrophobic membrane Size 1000mL Pack Description 1/Bag Case Quantity 12
Catalog No. 50-233-0209 Supplier Bellco Glass, Inc. Supplier No. 229584000
Available in two sizes 1000mL volume with 490cm2 of surface area and 2000mL volume with 850cm2 of surface area treated or nontreated versions. Expanded Surface Roller Bottle increases surface area for cell culture.Specifications Large knurled cap for easy grip Vented or nonvented cap for open or closed systems Manufactured from medical grade virgin polystyrene Tissue Culture Treated Roller Bottles have uniform hydrophilic surface ideal for consistent cell attachment Easytoread black printed graduations Vent caps feature 0.22µm hydrophobic membrane Size 1000mL Pack Description 1/Bag Case Quantity 12