Learn More
Thomas Scientific 2000mL Roller Bottle, Non-Treated Suspension Culture, Printed Graduations, Non-Vented Cap, Sterile
SDP
Catalog No. 501461509
Encompass_Preferred
Expanded Surface Roller Bottle (ESRB) 2000mL volume with ribbed surface providing 1900cm² of surface area Same dimensions as a standard 850cm 2 roller bottle, but fluted design provides 2X the surface area for cell culture so you can grow more cells without added cost of an additional bottle and media Roller Bottles 1000mL volume with 490cm 2 of surface area 2000mL volume with 850cm 2 of surface area 2000mL volume (ESRB) with 1900cm 2 of surface area Large knurled cap for easy grip Vented or non-vented caps for open or closed systems Manufactured from USP Class VI polystyrene Vacuum gas plasma tissue culture treated Uniform, hydrophilic surface facilitates cell attachment Easy-to-read black printed graduations Packaged sterile (gamma irradiated) in individual bags
Catalog No. 50-146-1509 Supplier Thomas Scientific Supplier No. 1201P92
Expanded Surface Roller Bottle (ESRB) 2000mL volume with ribbed surface providing 1900cm² of surface area Same dimensions as a standard 850cm 2 roller bottle, but fluted design provides 2X the surface area for cell culture so you can grow more cells without added cost of an additional bottle and media Roller Bottles 1000mL volume with 490cm 2 of surface area 2000mL volume with 850cm 2 of surface area 2000mL volume (ESRB) with 1900cm 2 of surface area Large knurled cap for easy grip Vented or non-vented caps for open or closed systems Manufactured from USP Class VI polystyrene Vacuum gas plasma tissue culture treated Uniform, hydrophilic surface facilitates cell attachment Easy-to-read black printed graduations Packaged sterile (gamma irradiated) in individual bags