Learn More
Chemglass Life Sciences Flask, Erlenmeyer, 2000mL, Sterile, Baffled, Polycarbonate, 53-B, with Blue Duo Cap, BULK PACK
SDP
Catalog No. 501532285
Encompass_Preferred
Flask, Erlenmeyer, 2000mL, Sterile, Baffled, Polycarbonate, 53-B, with Blue Duo Cap, BULK PACK. Quadruple baffled Erlenmeyer flasks are sterile, non-pyrogenic and manufactured with polycarbonate resin. Flasks are supplied with exclusive DuoCap designed for both aerobic and anaerobic applications (for aerobic applications the cap utilizes a 0.22m pore size PTFE membrane for sterile air exchange; for anaerobic applications the cap can easily be converted to a solid cap design). Solid top caps are also ideal for applications requiring mixing, shaking and storage of biological fluids. Graduations are molded for accuracy.EZ-Tear packaging allows for easy access.
Catalog No. 50-153-2285 Supplier Chemglass Life Sciences Supplier No. CLS2091200B
Flask, Erlenmeyer, 2000mL, Sterile, Baffled, Polycarbonate, 53-B, with Blue Duo Cap, BULK PACK. Quadruple baffled Erlenmeyer flasks are sterile, non-pyrogenic and manufactured with polycarbonate resin. Flasks are supplied with exclusive DuoCap designed for both aerobic and anaerobic applications (for aerobic applications the cap utilizes a 0.22m pore size PTFE membrane for sterile air exchange; for anaerobic applications the cap can easily be converted to a solid cap design). Solid top caps are also ideal for applications requiring mixing, shaking and storage of biological fluids. Graduations are molded for accuracy.EZ-Tear packaging allows for easy access.