Learn More
Ace Glass, Inc. 2000mL vacuum trap 160 x 410mm w/#25 thread
SDP
Catalog No. 50976308
Encompass_Preferred
2000mL vacuum trap with #25 Ace-Thred includes round bottom flask, adjustable inner tube, and nylon bushing with FETFE o-ring. Flask dimensions: 160mm x 410mm.
Catalog No. 50-976-308 Supplier Ace Glass, Inc. Supplier No. 874423
2000mL vacuum trap with #25 Ace-Thred includes round bottom flask, adjustable inner tube, and nylon bushing with FETFE o-ring. Flask dimensions: 160mm x 410mm.