Learn More
Electron Microscopy Sciences Ilford 2000rt Developer/Replenisher 5 LT
SDP
Catalog No. 5031377
Encompass
2000RT is a liquid concentrate developer/replenisher recommended for use with all types of black and white RC photo paper processors. It is easy to use as no starter solution is required. The recommended dilution for both the tank and replenisher solution is 1+4. It is a robust, economical, clean working developer with a long tank life. It can be used at a wide range of developing temperatures, 20 – 40 degree C (68 – 104 degree F). With most papers it gives a print image tone just slightly cool of neutral.
Catalog No. 50-313-77 Supplier Electron Microscopy Sciences Supplier No. 8305605
2000RT is a liquid concentrate developer/replenisher recommended for use with all types of black and white RC photo paper processors. It is easy to use as no starter solution is required. The recommended dilution for both the tank and replenisher solution is 1+4. It is a robust, economical, clean working developer with a long tank life. It can be used at a wide range of developing temperatures, 20 – 40 degree C (68 – 104 degree F). With most papers it gives a print image tone just slightly cool of neutral.