Learn More
Perkin Elmer US LLC 200µL, Buffer Tefzel Tubing
Catalog No. 502111658
Encompass_Preferred
This 200µL Tefzel (ETFE) Tubing delivers enhanced pressure-holding capabilities and continuous mechanical stability. Tefzel tubing is also ideal for solvent resistance.
Catalog No. 50-211-1658 Supplier Perkin Elmer US LLC Supplier No. N2992105
This 200µL Tefzel (ETFE) Tubing delivers enhanced pressure-holding capabilities and continuous mechanical stability. Tefzel tubing is also ideal for solvent resistance.