Learn More
Frontier Specialty Chemicals Bilirubin conjugate, 68683-34-1, MFCD00044931, 200MG
Supplier:  Frontier Specialty Chemicals B850200MG
Encompass_Preferred
Molecular Formula: C37 H44 N6 O10 S2 . 2 Na, Molecular Weight: 842.899. Bilirubin Conjugate is a water soluble ditaurate derivative of bilirubin equivalent to bilirubin glucuronide derivatives.
Catalog No. 50-981-733