Learn More
Avanti Polar Lipids 16:0 PC (DPPC), 63-89-8, 200mg
Supplier:  Avanti Polar Lipids 850355P200MG
Encompass
Molecular Formula C40H80NO8P, Purity >99%, Formula Weight 734.039, 1,2-dipalmitoyl-sn-glycero-3-phosphocholine
Catalog No. NC0212322