Learn More
U.S. Pharmacopeia Metformin hydrochloride, 1115-70-4, MFCD00012582, 200 mg
Catalog No. NC9365109
Encompass
Linear Formula: NH2C(=NH)NHC(=NH)N(CH3)2 · HCl, Molecular Weight: 165.62, MP: 223-226 °C, Synonym: 1,1-Dimethylbiguanide hydrochloride, Metformin.
Catalog No. NC9365109 Supplier U.S. Pharmacopeia Supplier No. 1396309
Linear Formula: NH2C(=NH)NHC(=NH)N(CH3)2 · HCl, Molecular Weight: 165.62, MP: 223-226 °C, Synonym: 1,1-Dimethylbiguanide hydrochloride, Metformin.