Learn More
Ace Glass, Inc. Flask, recovery, heavy wall, 200mL, 24/40
SDP
Catalog No. 509557456
Encompass_Preferred
200mL round bottom heavy wall recovery flask, with a single neck with a 24/40 joint, and a sloping side wall.
Catalog No. 50-955-7456 Supplier Ace Glass, Inc. Supplier No. 6892208
200mL round bottom heavy wall recovery flask, with a single neck with a 24/40 joint, and a sloping side wall.