Learn More
Chemglass Life Sciences Weighing Dish, Plastic, Hexagonal, Antistatic, 200mL, Polystyrene
SDP
Catalog No. 501215170
Encompass_Preferred
These dishes are excellent for handling solids or liquids during weighing. Dishes are easily bent into pouring spouts that enable non-spill transfer. The molded hexagonal design provides greater balance protection and safety. Not reactive with most substances. Dishes are suitable for weighing of static affected samples. They are stackable for easy storage and accessibility. Dishes will withstand temperatures up to 80°C.
Catalog No. 50-121-5170 Supplier Chemglass Life Sciences Supplier No. CLS1815009
These dishes are excellent for handling solids or liquids during weighing. Dishes are easily bent into pouring spouts that enable non-spill transfer. The molded hexagonal design provides greater balance protection and safety. Not reactive with most substances. Dishes are suitable for weighing of static affected samples. They are stackable for easy storage and accessibility. Dishes will withstand temperatures up to 80°C.