Learn More
Chemglass Life Sciences 200mL Filter Funnel, 24/40 Joint Size, 40mm OD Coarse Frit, Valve On Side Arm, Airfree, Schlenk
SDP
Catalog No. AF054224
Encompass_Preferred
200mL Filter Funnel, 24/40 Joint Size, 40mm OD Coarse Frit, Valve On Side Arm, Airfree, Schlenk. Fritted filter funnel has a choice of coarse or medium porosity fritted disc. Lower outlet is a standard taper inner joint and has a CG-961-01 0-4mm Chem-Vac™ Chem-Cap valve. Top outer joint is the same size as the lower inner joint. Capacity listed is that above the fritted disc. Available with a choice of a 2mm high vacuum glass stopcock having a Stainless Steel constant pressure retaining device or a 0-4mm Chem-Cap Airfree inlet valve.
Catalog No. AF054224 Supplier Chemglass Life Sciences Supplier No. AF054224
200mL Filter Funnel, 24/40 Joint Size, 40mm OD Coarse Frit, Valve On Side Arm, Airfree, Schlenk. Fritted filter funnel has a choice of coarse or medium porosity fritted disc. Lower outlet is a standard taper inner joint and has a CG-961-01 0-4mm Chem-Vac™ Chem-Cap valve. Top outer joint is the same size as the lower inner joint. Capacity listed is that above the fritted disc. Available with a choice of a 2mm high vacuum glass stopcock having a Stainless Steel constant pressure retaining device or a 0-4mm Chem-Cap Airfree inlet valve.