Learn More
Ace Glass, Inc. 200mL hemispherical fabric mantle, Glas-Col, 76mm max flask diameter, 1.75in inside depth, 4.50in OD, 2.50in height, 100 watts, CSA rated
SDP
Catalog No. 50977266
Encompass
200mL hemispherical fabric Glas-col mantle, adaptable to unusual shapes and sizes, and will withstand 450 Deg.C. Mantle has a separable 2-wire 4 cord with locking connector. cULus rated. CSA certified.
Catalog No. 50-977-266 Supplier Ace Glass, Inc. Supplier No. 1203511
200mL hemispherical fabric Glas-col mantle, adaptable to unusual shapes and sizes, and will withstand 450 Deg.C. Mantle has a separable 2-wire 4 cord with locking connector. cULus rated. CSA certified.