Learn More
Foxx Life Sciences PUREGRIP Rhino Safety Coated Glass Bottles, Amber, 2L, with Graduation Marks and GL45 VersaCap, 1/EA
GREENER_CHOICE
Catalog No. 502978695
Encompass_Preferred
PUREGRIP Rhino Bottles & Carboys excel in mixing, storing, and transporting media, buffers, chemicals, and solvents while enhancing lab safety. The Rhino Safety coating secures glass fragments if bottles break, minimizing risks, while its non-slip grip improves wet handling for a safer, more efficient lab. Made from durable 3.3 Borosilicate Glass and enabled with graduation marks to ensure precise measurements.
These products enhance lab safety through drip-free dispensing, minimizing spills, limiting chemical exposure, and enabling quick cleanup—an ideal choice for safety-focused labs.
Features
Shatter Resistant Coating
Non-Slip Exterior
Clear and Amber Options Available in sizes from 100mL to 20L
Thermal & Chemical Resistance: Withstands 121°C autoclaving and -20°C refrigeration
USP Class VI Certified: Includes an animal origin free polypropylene VersaCap with Secure Grasp technology for easy handling.
Catalog No. 50-297-8695 Supplier Foxx Life Sciences Supplier No. 48V9401FLS
PUREGRIP Rhino Bottles & Carboys excel in mixing, storing, and transporting media, buffers, chemicals, and solvents while enhancing lab safety. The Rhino Safety coating secures glass fragments if bottles break, minimizing risks, while its non-slip grip improves wet handling for a safer, more efficient lab. Made from durable 3.3 Borosilicate Glass and enabled with graduation marks to ensure precise measurements.
These products enhance lab safety through drip-free dispensing, minimizing spills, limiting chemical exposure, and enabling quick cleanup—an ideal choice for safety-focused labs.
Features
Shatter Resistant Coating
Non-Slip Exterior
Clear and Amber Options Available in sizes from 100mL to 20L
Thermal & Chemical Resistance: Withstands 121°C autoclaving and -20°C refrigeration
USP Class VI Certified: Includes an animal origin free polypropylene VersaCap with Secure Grasp technology for easy handling.