Learn More
Chemglass Life Sciences 200mL Glassblowers Pear Recovery Blank
SDP
Catalog No. CG62205
Encompass_Preferred
200mL Glassblowers Pear Recovery Blank. Blanks for fabrication of recovery flasks. NOTE: NECK IS UNFINISHED, FLASK CANNOT BE USED WITHOUT MODIFICATION OF NECK.
Catalog No. CG62205 Supplier Chemglass Life Sciences Supplier No. CG62205
200mL Glassblowers Pear Recovery Blank. Blanks for fabrication of recovery flasks. NOTE: NECK IS UNFINISHED, FLASK CANNOT BE USED WITHOUT MODIFICATION OF NECK.