Learn More
Ace Glass, Inc. Fabric heating mantle that covers top and bottom of 1, 2 or3 neck 200mL spherical flask. Lower half wattage is 100w at115 volts, CSA rated
SDP
Catalog No. 50977257
Encompass
Catalog No. 50-977-257 Supplier Ace Glass, Inc. Supplier No. 1203111