Learn More
Ace Glass, Inc. 200ML VESSEL 200ML VESSEL
SDP
Supplier:  Ace Glass, Inc. 669622
Encompass_Preferred
200mL vessel with sloped shoulders and a #25 Ace-Thred neck; includes PTFE lined cap. For use in freeze drying and storage. After drying material, vessel can be capped and stored. Working capacity of vessel is about 1/2 of volume.
Catalog No. 50-973-322