Learn More
Ace Glass, Inc. Condenser, Liebig, 200mm, 24/40
SDP
Catalog No. NC9874159
Encompass_Preferred
200mm Liebig condenser with 24/40 standard taper or spherical joint at bottom and top. Hose connections are size D.
Catalog No. NC9874159 Supplier Ace Glass, Inc. Supplier No. 599812
200mm Liebig condenser with 24/40 standard taper or spherical joint at bottom and top. Hose connections are size D.