Learn More
Chemglass Life Sciences Reaction Vessel Lid, 5-Neck, 45/50 Center Neck, 4-29/42 Side Necks, 200mm Schott Flange
SDP
Catalog No. CG194606
Encompass_Preferred
Reaction Vessel Lid, 5-Neck, 45/50 Center Neck, 4-29/42 Side Necks, 200mm Schott Flange. Five neck vessel lid to fit CG-1920 through CG-1936 vessels. Lid has a flat ground surface allowing it to be greased or used with an o-ring. Note: Five neck lids are available with joint sizes and configurations other than those listed, please call for details.
Catalog No. CG194606 Supplier Chemglass Life Sciences Supplier No. CG194606
Add to Cart
Add to Cart
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Reaction Vessel Lid, 5-Neck, 45/50 Center Neck, 4-29/42 Side Necks, 200mm Schott Flange. Five neck vessel lid to fit CG-1920 through CG-1936 vessels. Lid has a flat ground surface allowing it to be greased or used with an o-ring. Note: Five neck lids are available with joint sizes and configurations other than those listed, please call for details.