Learn More
Chemglass Life Sciences 200mm Quick Release Clamp, PTFE Coated
SDP
Catalog No. CG141T15
Encompass_Preferred
200mm Quick Release Clamp, PTFE Coated. Similar to CG-141 but with the entire clamp PTFE coated for chemical durability.
Catalog No. CG141T15 Supplier Chemglass Life Sciences Supplier No. CG141T15
200mm Quick Release Clamp, PTFE Coated. Similar to CG-141 but with the entire clamp PTFE coated for chemical durability.